Product Name
N-Phenyl-o-phenylenediamine 98%
Product Number
P28352
Product Brand
ALDRICH
CAS Number
534-85-0
Molecular Weight
184.24
Test | Specification |
---|---|
APPEARANCE | PINK, PURPLE OR BROWN POWDER, CRYSTALS, |
CRYSTALLINE POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5%-102.5% (WITH HCLO4) |
OR | |
ELEMENTAL ANALYSIS | CARBON 76.3%-80.2% |
NITROGEN 14.8%-15.6% | |
AND/OR | |
GAS LIQUID | 97.5% (MINIMUM) |
APPROVED JULY 18, 2005 RJM |