Product Name
6-Aminohexanoic acid ≥98.5% (NT)
Product Number
07260
Product Brand
SIAL
CAS Number
60-32-2
Molecular Weight
131.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION (NT) HCLO4 0.1M | 98.5 - 101.0 % |
SOLUBILITY (COLOR) | REPORT RESULT |
SOLUBILITY (TURBIDITY) | REPORT RESULT |
SOLUBILITY (METHOD) | 1G IN 10ML WATER |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |