Product Name
Ammonium oxalate monohydrate BioUltra, ≥99.5% (RT)
Product Number
09898
Product Brand
SIAL
CAS Number
6009-70-7
Molecular Weight
142.11
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals |
REDOX TITRATION | 99.5 - 101.0 % |
REDOXTITRATION (METHOD) | PERMANGANATE TITRATION |
RESIDUE ON IGNITION | ≤ 0.02 % |
METAL TRACE ANALYSIS (ICP) | CORRESPONDS TO REQUIREMENTS |
ALUMINIUM (ICP) | ≤ 5 mg/kg |
BARIUM (ICP) | ≤ 5 mg/kg |
BISMUTH (ICP) | ≤ 5 mg/kg |
CALCIUM (ICP) | ≤ 10 mg/kg |
CADMIUM (ICP) | ≤ 5 mg/kg |
COBALT (ICP) | ≤ 5 mg/kg |
CHROMIUM (ICP) | ≤ 5 mg/kg |
COPPER (ICP) | ≤ 5 mg/kg |
IRON (ICP) | ≤ 2 mg/kg |
POTASSIUM (ICP) | ≤ 50 mg/kg |
LITHIUM (ICP) | ≤ 5 mg/kg |
MAGNESIUM (ICP) | ≤ 5 mg/kg |
MANGANESE (ICP) | ≤ 5 mg/kg |
MOLYBDENUM (ICP) | ≤ 5 mg/kg |
SODIUM (ICP) | ≤ 50 mg/kg |
NICKEL (ICP) | ≤ 10 mg/kg |
LEAD (ICP) | ≤ 5 mg/kg |
STRONTIUM (ICP) | ≤ 5 mg/kg |
ZINC (ICP) | ≤ 5 mg/kg |
ARSENIC TRACES (MHS-AAS) | ≤ 0.1 mg/kg |
TOTAL SULFUR AS SO4 (ICP) | ≤ 20 mg/kg |
CHLORIDE (CL) | ≤ 5 mg/kg |
SOLUBILITY (METHOD) | 0.1M water |
APPEARANCE (SOLUTION) | CLEAR COLORLESS |
PH (SOLUTION) | 6.0 - 7.0 |
RESIDUE (FILTER TEST) | NO RESIDUE |
CUT-OFF | 0 - 267 nm |
RECOMMENDED RETEST PERIOD | 72 MONTHS |