Product Name
Benzoic acid ReagentPlus®, 99%
Product Number
109479
Product Brand
SIAL
CAS Number
65-85-0
Molecular Weight
122.12
Test | Specification |
---|---|
SOLUTION | |
REVISED FEBRUARY 3, 2006 RJM | |
APPEARANCE | WHITE CHIPS OR POWDER |
GAS LIQUID | 98.5% (MINIMUM) |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
SOLUBILITY | 10% IN 95% ETHANOL; CLEAR, COLORLESS |
TITRATION | 98.5% - 101.5% (WITH NAOH) |