Product Name
Succinonitrile purum, ≥97.0% (GC)
Product Number
14190
Product Brand
SIAL
CAS Number
110-61-2
Molecular Weight
80.09
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow |
APPEARANCE (FORM) | Powder or Solid or Crystals |
PURITY (GC AREA %) | ≥ 97.0 % |
CARBON CONTENT | 59.99 % (THEORY) |
HYDROGEN CONTENT | 5.03 % (THEORY) |
NITROGEN CONTENT | 34.98 % (THEORY) |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |