Product Name
4-Nitroaniline analytical standard
Product Number
31569
Product Brand
SIAL
CAS Number
100-01-6
Molecular Weight
138.12
Test | Specification |
---|---|
APPEARANCE (COLOR) | Light Beige to Very Dark Beige and Light Brown to Very Dark Brown |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 98.0 % |
MELTING POINT | 145 - 150 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |