Product Name
2,6-TDI analytical standard
Product Number
33493
Product Brand
SIAL
CAS Number
91-08-7
Molecular Weight
174.16
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow to Light Brown-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 98.0 % |
REFRACTIVE INDEX N20/D | 1.5695 - 1.5735 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 48 MONTHS |