Product Name
Foramsulfuron PESTANAL®, analytical standard
Product Number
33977
Product Brand
SIAL
CAS Number
173159-57-4
Molecular Weight
452.44
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
WATER | ≤ 5.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 48 MONTHS |