Product Name
Avenanthramide C analytical standard
Product Number
36465
Product Brand
SIAL
CAS Number
116764-15-9
Molecular Weight
315.28
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Dark Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
IDENTIFICATION (LC-MS) | CORRESPONDS TO REQUIREMENTS |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 24 MONTHS |