Product Name
2,5-Dichloroaniline PESTANAL®, analytical standard
Product Number
36702
Product Brand
SIAL
CAS Number
95-82-9
Molecular Weight
162.02
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Yellow and Faint Beige to Beige to Brown and Faint Grey to Light Grey |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 98.0 % |
MELTING POINT | 46 - 52 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |