Product Name
Glufosinate-ammonium PESTANAL®, analytical standard
Product Number
45520
Product Brand
SIAL
CAS Number
77182-82-2
Molecular Weight
198.16
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
MELTING POINT | 206 - 216 C |
WATER | ≤ 2.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 48 MONTHS |