Product Name
Pendimethalin solution 100 μg/mL in methanol, PESTANAL®, analytical standard
Product Number
45892
Product Brand
SIAL
CAS Number
40487-42-1
Molecular Weight
281.31
Test | Specification |
---|---|
APPEARANCE (COLOR) | Light Yellow to Very Dark Yellow and Light Green-Yellow to Very Dark Green-Yellow |
APPEARANCE (FORM) | Liquid |
CONCENTRATION COMP1 | ~ 100 ug/ml |
SOLVENT | METHANOL |
PURITY (HPLC AREA %) | ≥ 98.0 % |
IDENTIFICATION (LC-MS) | Identity Confirmed |
REFRACTIVE INDEX N20/D | 1.3266 - 1.3306 |
EXPIRY PERIOD | 60 MONTHS |