Product Name
trans-2-Octenal analytical standard
Product Number
52464
Product Brand
SIAL
CAS Number
2548-87-0
Molecular Weight
126.20
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 97.0 % |
REFRACTIVE INDEX N20/D | 1.450 - 1.455 |
WATER (COULOMETR.) | ≤ 0.5 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |