Product Name
Iohexol analytical standard
Product Number
74147
Product Brand
SIAL
CAS Number
66108-95-0
Molecular Weight
821.14
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0% (sum of isomers) |
WATER | ≤ 6.0 % |
EXPIRY PERIOD | 36 MONTHS |