Product Name
Sodium 1-nonanesulfonate suitable for ion pair chromatography, LiChropur™, ≥99.0% (T)
Product Number
74316
Product Brand
SIAL
CAS Number
35192-74-6
Molecular Weight
230.30
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Flakes |
TITRATION (ION EXCHANGE) | ≥ 99.0 % |
SUITABILITY TEST (HPLC) | CORRESPONDS TO STANDARD |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
UV Absorbance 210nm | ≤ 0.05 |
UV Absorbance 220nm | ≤ 0.03 |
UV Absorbance 230nm | ≤ 0.02 |
UV Absorbance 260nm | ≤ 0.02 |
UV Absorbance 500nm | ≤ 0.02 |
CYCLIC VOLTAMMOGRAM | CORRESPONDS TO STANDARD CV |
RESIDUE (FILTER TEST) | NO RESIDUE |
RECOMMENDED RETEST PERIOD | 42 MONTHS |