Product Name
N1,N12-Diacetylspermine hydrochloride analytical standard
Product Number
91423
Product Brand
SIAL
CAS Number
61345-83-3
Molecular Weight
286.41 (free base basis)
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
CARBON CONTENT | 42.48 % (THEORY, CALC. FOR TRIHYDROCHLORIDE) |
HYDROGEN CONTENT | 8.40 % (THEORY, CALC. FOR TRIHYDROCHLORIDE) |
NITROGEN CONTENT | 14.16 % (THEORY, CALC. FOR TRIHYDROCHLORIDE) |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |