Product Name
p-Toluenesulfonyl chloride reagent grade, ≥98%
Product Number
T35955
Product Brand
SIAL
CAS Number
98-59-9
Molecular Weight
190.65
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
ARGENTOMETR. TITRATION | 98.0 - 102.0 % |
PURITY (GC AREA %) | ≥ 98.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |