Product Name
Perchloric acid puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Product Number
77234
Product Brand
SIGALD
CAS Number
7601-90-3
Molecular Weight
100.46
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
COLOR (IN APHA) | ≤ 10 APHA |
TITRATION (T) NAOH 1M | 60.0 - 62.0 % |
DENSITY D20/4 | 1.536 - 1.558 |
SULFATED ASH | ≤ 30 mg/kg |
Free chlorine | ≤ 0.5 mg/kg |
HEAVY METALS | ≤ 0.0001 % |
Miscellaneous Assay | ≤ 5 mg/kg Posphate, Silicate (as SiO2) |
Miscellaneous Assay | ≤ 10 mg/kg Chlorate (ClO3) |
ACS SPECIFICATIONS | MEETS CURRENT ACS REQUIREMENTS |
SILVER (ICP) | ≤ 0.1 mg/kg |
ALUMINIUM (ICP) | ≤ 0.05 mg/kg |
ARSENIC (ICP) | ≤ 0.05 mg/kg |
BARIUM (ICP) | ≤ 0.02 mg/kg |
BISMUTH (ICP) | ≤ 0.1 mg/kg |
CALCIUM (ICP) | ≤ 0.5 mg/kg |
CADMIUM (ICP) | ≤ 0.05 mg/kg |
COBALT (ICP) | ≤ 0.05 mg/kg |
COPPER (ICP) | ≤ 0.1 mg/kg |
IRON (ICP) | ≤ 1 mg/kg |
POTASSIUM (ICP) | ≤ 0.1 mg/kg |
LITHIUM (ICP) | ≤ 0.02 mg/kg |
MAGNESIUM (ICP) | ≤ 0.5 mg/kg |
MANGANESE (ICP) | ≤ 0.02 mg/kg |
MOLYBDENUM (ICP) | ≤ 0.05 mg/kg |
NICKEL (ICP) | ≤ 0.1 mg/kg |
LEAD (ICP) | ≤ 0.05 mg/kg |
STRONTIUM (ICP) | ≤ 0.02 mg/kg |
THALLIUM (ICP) | ≤ 0.05 mg/kg |
ZINC (ICP) | ≤ 0.1 mg/kg |
METAL TRACE ANALYSIS (ICP-MS) | CORRESPONDS TO REQUIREMENTS |
CHLORIDE (CL) | ≤ 3 mg/kg |
TOTAL NITROGEN | ≤ 10 mg/kg |
SULFATE (SO4) | ≤ 10 mg/kg |
RECOMMENDED RETEST PERIOD | 6 YEARS |