Product Name
Quin-2 suitable for fluorescence, ≥95% (HPLC)
Product Number
08520
Product Brand
SIGMA
CAS Number
73630-23-6
Molecular Weight
693.87
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
LUMINESCENCE | CORRESPONDS TO REQUIREMENTS |
EMISSION MAXIMUM | 491.0 - 501.0 nm |
RECOMMENDED RETEST PERIOD | 24 MONTHS |