Product Name
Aminoallyl-dUTP-Cy®5 triethylammonium salt solution ≥95.0% (HPLC)
Product Number
19475
Product Brand
SIGMA
CAS Number
Molecular Weight
1162.01 (free acid basis)
Test | Specification |
---|---|
APPEARANCE (COLOR) | Blue |
APPEARANCE (FORM) | Liquid |
PURITY (HPLC AREA %) | ≥ 95.0 % |
IDENTIFICATION (LC-MS) | CORRESPONDS TO REQUIREMENTS |
PH-TEST | 7.0 - 8.0 |
RECOMMENDED RETEST PERIOD | 12 MONTHS |