Product Name
4-Nitrophenyl octanoate ≥90.0% (GC)
Product Number
21742
Product Brand
SIGMA
CAS Number
1956-10-1
Molecular Weight
265.30
Test | Specification |
---|---|
APPEARANCE (COLOR) | Light Green-Yellow to Very Dark Green-Yellow to Very Dark Brown-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 90.0 % |
REFRACTIVE INDEX N20/D | 1.504 - 1.512 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 36 MONTHS |