Product Name
Abberior® STAR RED NHS ≥80.0% (degree of coupling)
Product Number
43354
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Blue to Very Dark Blue |
APPEARANCE (FORM) | Powder or Crystals |
SOLUBILITY (COLOR) | for information |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 1 mg/ml in water |
DEGREE OF COUPLING | ≥ 80.0 % |
EMISSION MAXIMUM | 660 ± 10 nm |
REMARKS ON LUMINESCENCE | Ex. 637 nm in water |
RECOMMENDED RETEST PERIOD | 24 MONTHS |