Product Name
Methyl cellulose medium viscosity, Methoxyl content 27.5-31.5 %
Product Number
64625
Product Brand
SIGMA
CAS Number
9004-67-5
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder |
LOSS ON DRYING | ≤ 5.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
STRUCTURAL VISCOSITY | 1200 - 1800 mPas |
VISCOSITY (CONDITIONS) | 20 C , 2 % IN H2O / DIN 45, 10S-1 |
Miscellaneous Assay | METHOXYL CONTENT: 27.5 - 31.5 % |