Product Name
Methoxatin disodium salt ≥97.0% (HPLC)
Product Number
80198
Product Brand
SIGMA
CAS Number
122628-50-6
Molecular Weight
374.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | RED-BROWN TO DEEP RED-BROWN OR DEEP ORANGE |
APPEARANCE (FORM) | Powder |
PURITY (HPLC AREA %) | ≥ 97.0 % |
CARBON CONTENT | 44.94 % (theory) |
HYDROGEN CONTENT | 1.08 % (theory) |
NITROGEN CONTENT | 7.49 % (theory) |
REM. ON ELEMENTAL ANALYSIS | Water: 10 - 15 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |