Product Name
Glyceryl trioleate ≥97.0% (TLC)
Product Number
92860
Product Brand
SIGMA
CAS Number
122-32-7
Molecular Weight
885.43
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Faint Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 97.0 % |
PURITY (TLC AREA %) | ≥ 97.0 % |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 0.1G/ML CHLOROFORM |