Product Name
Ala-Leu
Product Number
A1878
Product Brand
SIGMA
CAS Number
3303-34-2
Molecular Weight
202.25
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS TO VERY SLIGHTLY HAZY SOLUTION AT 50 MG/ML IN WATER |
WATER CONTENT BY KARL FISCHER | < OR = 8.0% |
CARBON | 49.1% TO 54.5 |
NITROGEN | 12.7% TO 14.1 |
PURITY BY THIN LAYER CHROMATOGRAPHY | >99% |
DOCUMENT # | A1878/05/15/08/1 |