Product Name
Monoclonal Anti-APOL1 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL0173, purified immunoglobulin, buffered aqueous glycerol solution
Product Number
AMAB90532
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report Result |
Western Blot Dilution | 1:500 - 1:1000 |