Product Name
Monoclonal Anti-FCGRT antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL3640, purified immunoglobulin, buffered aqueous solution
Product Number
AMAB91200
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report Result |
Western Blot Dilution | 1:500 - 1:1000 |
Immunohistochemistry Dilution | 1:5000 - 1:10000 |
Immunohistochemistry Antibody Retrieval Method | HIER pH6 |