Product Name
Monoclonal Anti-TP63 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL3748, purified immunoglobulin, buffered aqueous glycerol solution
Product Number
AMAB91224
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | Report Result |
Western Blot Dilution | 1:500 - 1:1000 |
Immunohistochemistry Dilution | 1:1000 - 1:2500 |
Immunohistochemistry Antibody Retrieval Method | HIER pH6 |