Product Name
Bradykinin Fragment 1-5 ≥97% (HPLC)
Product Number
B1401
Product Brand
SIGMA
CAS Number
23815-89-6
Molecular Weight
572.66
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS SOLUTION AT 1 MG/ML IN WATER |
PEPTIDE CONTENT BY AMINO ACID ANALYSIS | > OR = 60% |
PURITY BY HPLC | > OR = 97% |
DOCUMENT # | B1401/05/07/08/1 |