Product Name
Sodium 1-butanesulfonate anionic detergent
Product Number
B2013
Product Brand
SIGMA
CAS Number
2386-54-1
Molecular Weight
160.17
Test | Specification |
---|---|
APPEARANCE | WHITE POWDER |
SOLUBILITY | CLEAR TO SLIGHTLY HAZY COLORLESS SOLUTION AT 100 MG/ML IN WATER |
IR SPECTRUM | CONSISTENT WITH STRUCTURE |
CARBON | 29.4-30.4% |
B2013/10/16/06 |