Product Name
Boc-Val-Pro-Arg-7-amido-4-methylcoumarin hydrochloride
Product Number
B9385
Product Brand
SIGMA
CAS Number
70375-24-5
Molecular Weight
627.73
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS SOLUTION AT 20MG/ML IN WATER |
IR SPECTRUM | CONSISTENT WITH STRUCTURE |
WATER BY KARL FISCHER | NMT 5.4% (2MOL/MOL) |
PURITY BY THIN LAYER CHROMATOGRAPHY | NLT 98% |
NOTE | FORMULA AND FORMULA WEIGHT ARE FOR THE FREE BASE |
DOCUMENT # B9385/09/29/00/2 |