Product Name
MISSION® esiRNA targeting mouse Ucp2
Product Number
EMU024521
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | 200 ng/µL |
PCR Product - Gel Electrophoresis | Pass |
PCR Product - DNA Sequencing | Pass |
Concentration - OD260 or Fluorescence Intensity | Pass |
Purified RNaseIII - Gel Electrophoresis | Pass |