Product Name
Anti-ACY1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Number
HPA036175
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Protein Array | pass |
Immunohistochemistry Dilution | 1:200 - 1:500 |
Immunohistochemistry Antibody Retrieval Method | HIER pH6 |
Concentration | 0.05 mg/mL |