Product Name
Porcine Serum USA origin, sterile-filtered, suitable for cell culture
Product Number
P9783
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
COUNTRY OF ORIGIN | UNITED STATES |
COUNTRY OF PROCESSING | UNITED STATES |
APPEARANCE | CLEAR STRAW TO AMBER LIQUID |
PH TEST | 7.2 - 8.0 |
OSMOLALITY | 280 - 345 MOSM/KG H2O |
STERILITY BY USP | NEGATIVE |
MYCOPLASMA TEST BY BARILE | NONE DETECTED |
HEMOGLOBIN | NMT 50 MG% |
ENDOTOXIN ASSAY | NMT 20.0 EU/ML |
CHOLESTEROL | REPORT RESULT |
TRIGLYCERIDES | REPORT RESULT |
GLUCOSE | REPORT RESULT |
SODIUM | REPORT RESULT |
IRON | REPORT RESULT |
PROTEIN CONTENT | 7.0 - 9.5 G% |
ALBUMIN | REPORT RESULT |
ALPHA GLOBULINS | REPORT RESULT |
BETA GLOBULINS | REPORT RESULT |
GAMMA GLOBULINS | REPORT RESULT |
MYCOPLASMA HYOPNEUMONIA BY ELISA | REPORT RESULT ON RAW MATERIAL |
PORCINE VIRUS 9CFR | NONE DETECTED ON RAW MATERIAL FOR BVD, TGE, PPV, REOVIRUS, RABIES, PAV, HEV, AND PRRS |
SWINE INFLUENZA VIRUS HEMAGGLUTINATION INHIBITION | REPORT RESULT ON RAW MATERIAL |
CELL CULTURE TEST | PASS |
EXPIRATION DATE | 48 MONTHS |
P9783:12/06-9 |