Product Name
Anti-P53R2 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
PRS2383
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting | p53R2 antibody can be used for detection of p53R2 by Western blot at 0. 5 to 1 ug/mg/ml. A 39 kDa band can be detected. |
ELISA | P53R2 antibody can be used for detection of 10 ng of immunogenic P53R2 peptide by ELISA at 0.01 - 1 ug/ml. |
Immunohistochemistry | P53R2 antibody can be used for immunohistochemistry at at 1 ug/ml. |
Immunofluorescence | Unverified |
Immunofluorescence | Unverified |