Product Name
Anti-VISA (ab2) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
PRS4055
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting | VISA antibody can be used for detection of VISA by Western blot at 0. 5 � 1 ug/mlg/ml. |
ELISA | VISA antibody can be used for detection of 10 ng of immunogenic VISA peptide by ELISA at 0.01 - 1 ug/ml. |
Immunohistochemistry | VISA antibody can be used for immunohistochemistry at at 2.5 ug/ml. |
Immunofluorescence | Unverified |
Immunofluorescence | Unverified |