Product Name
Anti-GDF6 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
PRS4691
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting | GDF6 antibody can be used for detection of GDF6 by Western blot at 1 ug/mlg/ml. |
ELISA | GDF6 antibody can be used for detection of 10 ng of immunogenic GDF6 peptide by ELISA at 0.01 - 1 ug/ml. |
Immunohistochemistry | Unverified |
Immunofluorescence | Unverified |
Immunofluorescence | Unverified |