Product Name
SGK2 Active human ≥75% (SDS-PAGE), aqueous glycerol solution, recombinant, expressed in E. coli
Product Number
S7570
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
PROTEIN BY BRADFORD | REPORT RESULT |
ENZYMATIC ACTIVITY | MINIMUM 100 UNITS/MG PROTEIN |
UNIT DEFINITION | ONE UNIT WILL INCORPORATE 1NANOMOLE OF PHOSPHATE INTO THE AKT/SGK SUBSTRATE PEPTIDE (RPRAATF) PER MINUTE AT PH7.2 AT 30DEGC USING A FINAL CONCENTRATION OF 50UM (32P)ATP. |
PURITY BY SDS-PAGE | NLT 75% |
DOCUMENT # S7570/06/13/05/1 |