Product Name
ANTI-MYOGENIN(N-TERMINAL) antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Product Number
SAB1305721
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report Result |
Concentration | Report Result |
Immunohistochemistry | Not determined |
Western Blotting ( Cell Lysate) | 1:1000 |
Flow Cytometry | Not determined |
ELISA | Not determined |
Immunoprecipitation | Not determined |
Chromatin Immunoprecipitation | Not determined |
Immunofluorescence | Not determined |