Product Name
Monoclonal Anti-IFNA13 antibody produced in mouse clone 3F9, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1401198
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~37 kDa using immunogen protein lysate. |