Product Name
Anti-SUMO2 antibody produced in rabbit purified immunoglobulin, buffered aqueous solution
Product Number
SAB1401366
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~10.90 kDa using SUMO2 transfected 293T lysate. |
Detection Antibody | Suitable as a detection antibody in sandwich ELISA assay. |