Product Name
Anti-TFAM antibody produced in mouse purified immunoglobulin, buffered aqueous solution
Product Number
SAB1401382
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Tissue lysate) | Antibody concentration: 1 - 5 ug/ml. Specific band of ~29.1 kDa using human kidney lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 3 ug/ml using human small Intestine tissue. |
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~27.06 kDa using TFAM transfected 293T lysate. |
Detection Antibody | Suitable as a detection antibody in sandwich ELISA assay. |