Product Name
Anti-SOX18 affinity isolated antibody
Product Number
SAB2108480
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | 1 mg/ml |
ELISA | Not Verified |
Western Blotting | Antibody dilution: 0.5 µg/ml |
Immunohistochemistry | Conditions: Paraffin-embedded tissue. Antibody Concentration: 4-8µg/ml. |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |