Product Name
Anti-PALS/MPP5 antibody produced in goat affinity isolated antibody, buffered aqueous solution
Product Number
SAB2500743
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | 100µg/200µL |
ELISA (Peptide) | Antibody detection limit dilution 1:32000. |
Western Blotting (Cell lysate) | Approx. 75kDa band observed in Human Brain lysate. Recommended concentration: 1-3 µg/ml. |
Immunohistochemistry - Paraffin Embedded Tissue | In paraffin embedded Human Skeletal Muscle shows strong alternating staning of muscle fibres in longitudinal section. Recommended concentration: 5-10µg/ml. |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |