Product Name
Anti-PTPN6/SHP1 (Internal) antibody produced in goat affinity isolated antibody, buffered aqueous solution
Product Number
SAB2500840
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless liquid |
Concentration | 100µg/200µL |
ELISA (Peptide) | Antibody detection limit dilution 1:128000. |
Western Blotting (Cell lysate) | Approx. 70kDa band observed in lysates of cell lines Jurkat and U937. Recommended concentration: 0.2-0.5 µg/ml. |
Immunohistochemistry - Paraffin Embedded Tissue | Not Verified |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |