Product Name
Anti-AP1G1 antibody produced in goat affinity isolated antibody, buffered aqueous solution
Product Number
SAB2900324
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Liquid |
Concentration | 1 mg/ml |
ELISA | Antibody concentration: 1:2000-1:8000 |
Western Blotting | Antibody concentration: 1:500-1:2000 |
Immunohistochemistry | Antibody concentration: 2.5 µg/ml |
Immunofluorescence | Not Verified |
Immunoprecipitation | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |