Product Name
Monoclonal Anti-PD-1 antibody produced in mouse clone 7A11B1, purified immunoglobulin, buffered aqueous solution
Product Number
SAB3500122
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Concentration | 1 mg/mL |
ELISA | 0.01-1 ug/mL |
Western Blotting | 1 ug/mL |
Immunohistochemistry | 2.5 ug/mL |
Immunocytochemistry | Not Verified |
Flow Cytometry | Not Verified |
Chromatin Immunoprecipitation | Not Verified |