Product Name
Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in donkey affinity isolated antibody, lyophilized powder
Product Number
SAB3700934
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
ELISA | 1:40,000 |
Western blotting | 1:1,000 - 1:3,000 |
Immunohistochemistry | 1:500 - 1:2,000 |