Product Name
Anti-phospho-PEK/PERK (pThr981) antibody produced in rabbit affinity isolated antibody
Product Number
SAB4301310
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Liquid |
Concentration | 1.0mg/mL |
ELISA | Not Determined |
Sandwhich ELISA | Not Determined |
Western Blotting (Cell Lysate) | Not Determined |
Immunohistochemistry | 1:50~1:100 |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytopmetry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |